Assay Details
| Gene Symbol: HIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198259.1 | NP_001185188.1 | 9 - 10 | 1039 | 89 | |
| NM_001333440.1 | NP_001322424.1 | 8 - 9 | 1269 | 89 | ||
| NM_001333441.1 | NP_001322422.1 | 9 - 10 | 1068 | 89 | ||
| NM_001333442.1 | NP_001322423.1 | 9 - 10 | 1024 | 89 | ||
| NM_103933.6 | NP_564573.3 | 9 - 10 | 1165 | 89 | ||
| GenBank mRNA | AK228745.1 | - | 9 - 10 | 1020 | 89 | |
More Information
Set Membership: |
|

HIT1
Genomic Map