Assay Details
| Gene Symbol: AT1G50575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103939.4 | NP_564577.1 | 3 - 4 | 735 | 120 | |
| GenBank mRNA | AY059883.1 | - | 3 - 4 | 624 | 120 | |
| AY093371.1 | - | 3 - 4 | 533 | 120 | ||
More Information
Set Membership: |
|

AT1G50575
Genomic Map