Assay Details
| Gene Symbol: HAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085089.1 | NP_001078558.1 | 1 - 2 | 354 | 79 | |
| NM_121011.4 | NP_196536.1 | 1 - 2 | 361 | 79 | ||
| GenBank mRNA | AB493744.1 | - | 1 - 2 | 309 | 79 | |
More Information
Set Membership: |
|

HAM2
Genomic Map