Assay Details
| Gene Symbol: HAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085089.1 | NP_001078558.1 | 5 - 6 | 713 | 77 | |
| NM_121011.4 | NP_196536.1 | 6 - 7 | 954 | 77 | ||
| GenBank mRNA | AB493744.1 | - | 6 - 7 | 902 | 77 | |
More Information
Set Membership: |
|

HAM2
Genomic Map