Assay Details
| Gene Symbol: AT5G09800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121017.3 | NP_196542.1 | 1 | 1245 | 117 | |
| GenBank mRNA | AK175195.1 | - | 1 | 1112 | 117 | |
| AK176549.1 | - | 1 | 1112 | 117 | ||
| BT010883.1 | - | 1 | 1051 | 117 | ||
More Information
Set Membership: |
|

AT5G09800
Genomic Map