Assay Details
| Gene Symbol: THO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121023.4 | NP_568219.1 | 13 - 14 | 1217 | 96 | |
| GenBank mRNA | AF424566.1 | - | 13 - 14 | 1137 | 96 | |
| AF428323.1 | - | 13 - 14 | 1155 | 96 | ||
More Information
Set Membership: |
|

THO1
Genomic Map