Assay Details
| Gene Symbol: THO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203340.2 | NP_001190269.1 | 8 - 9 | 877 | 154 | |
| NM_121023.4 | NP_568219.1 | 8 - 9 | 861 | 154 | ||
| GenBank mRNA | AF424566.1 | - | 8 - 9 | 781 | 154 | |
| AF428323.1 | - | 8 - 9 | 799 | 154 | ||
More Information
Set Membership: |
|

THO1
Genomic Map