Assay Details
| Gene Symbol: CESA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121024.3 | NP_196549.1 | 5 - 6 | 1208 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CESA5| Gene Symbol: CESA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121024.3 | NP_196549.1 | 5 - 6 | 1208 | 85 | |
Set Membership: |
|