Assay Details
| Gene Symbol: AT5G09880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121025.4 | NP_568220.1 | 1 - 2 | 561 | 65 | |
| GenBank mRNA | AY054603.1 | - | 1 - 2 | 524 | 65 | |
More Information
Set Membership: |
|

AT5G09880
Genomic Map