Assay Details
| Gene Symbol: AT5G09880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121025.4 | NP_568220.1 | 4 - 5 | 744 | 87 | |
| GenBank mRNA | AY054603.1 | - | 4 - 5 | 707 | 87 | |
More Information
Set Membership: |
|

AT5G09880
Genomic Map