Assay Details
| Gene Symbol: AT5G09890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121026.5 | NP_568221.1 | 10 - 11 | 1712 | 103 | |
| GenBank mRNA | AB047278.1 | - | 10 - 11 | 1513 | 103 | |
| AY034932.1 | - | 10 - 11 | 1576 | 103 | ||
| AY059113.1 | - | 10 - 11 | 1435 | 103 | ||
More Information
Set Membership: |
|

AT5G09890
Genomic Map