Assay Details
| Gene Symbol: AT5G09890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036786.1 | NP_001031863.1 | 7 - 8 | 1365 | 84 | |
| NM_121026.5 | NP_568221.1 | 7 - 8 | 1474 | 84 | ||
| GenBank mRNA | AB047278.1 | - | 7 - 8 | 1275 | 84 | |
| AY034932.1 | - | 7 - 8 | 1338 | 84 | ||
| AY059113.1 | - | 7 - 8 | 1197 | 84 | ||
More Information
Set Membership: |
|

AT5G09890
Genomic Map