Assay Details
| Gene Symbol: AT5G09910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343075.1 | NP_001318521.1 | 6 - 7 | 1087 | 97 | |
| NM_001343076.1 | NP_001330738.1 | 6 - 7 | 938 | 97 | ||
| NM_001343077.1 | NP_001330737.1 | 6 - 7 | 922 | 97 | ||
| NM_001343078.1 | NP_001330736.1 | 5 - 6 | 631 | 97 | ||
| NM_121028.4 | NP_196553.2 | 5 - 6 | 1004 | 97 | ||
More Information
Set Membership: |
|

AT5G09910
Genomic Map