Assay Details
| Gene Symbol: NRPB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121029.3 | NP_196554.1 | 4 - 5 | 359 | 92 | |
| GenBank mRNA | AF016511.1 | - | 4 - 5 | 306 | 92 | |
| AK317113.1 | - | 4 - 5 | 359 | 92 | ||
| BT025500.1 | - | 4 - 5 | 266 | 92 | ||
More Information
Set Membership: |
|

NRPB4
Genomic Map