Assay Details
| Gene Symbol: AT5G09960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343081.1 | NP_001332505.1 | 2 - 3 | 932 | 66 | |
| NM_001343082.1 | NP_001332506.1 | 2 - 3 | 964 | 66 | ||
| NM_121033.3 | NP_568222.1 | 2 - 3 | 489 | 66 | ||
| GenBank mRNA | AK176727.1 | - | 2 - 3 | 379 | 66 | |
| BT024549.1 | - | 2 - 3 | 288 | 66 | ||
More Information
Set Membership: |
|

AT5G09960
Genomic Map