Assay Details
| Gene Symbol: PROPEP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121036.3 | NP_568224.1 | 1 - 2 | 194 | 150 | |
| GenBank mRNA | AK176126.1 | - | 1 - 2 | 193 | 150 | |
| AY086181.1 | - | 1 - 2 | 194 | 150 | ||
| BT024633.1 | - | 1 - 2 | 139 | 150 | ||
More Information
Set Membership: |
|

PROPEP5
Genomic Map