Assay Details
| Gene Symbol: AT5G10420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121080.3 | NP_196604.2 | 1 - 2 | 322 | 73 | |
| GenBank mRNA | DQ446939.1 | - | 1 - 2 | 277 | 73 | |
More Information
Set Membership: |
|

AT5G10420
Genomic Map