Assay Details
| Gene Symbol: RBK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121090.4 | NP_568231.1 | 6 - 7 | 1363 | 70 | |
| GenBank mRNA | AY150507.1 | - | 5 - 6 | 1102 | 70 | |
More Information
Set Membership: |
|

RBK1
Genomic Map