Assay Details
| Gene Symbol: AT5G10530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121091.2 | NP_196615.1 | 1 | 2842 | 93 | |
| GenBank mRNA | DQ446940.1 | - | 1 | 1907 | 93 | |
| Gene Symbol: AT5G10540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121092.4 | NP_568232.1 | 1 | 216 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_121092.4 transcripts.
Set Membership: |
|

AT5G10530
Genomic Map