Assay Details
| Gene Symbol: AT5G10540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121092.4 | NP_568232.1 | 17 - 18 | 6038 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G10540| Gene Symbol: AT5G10540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121092.4 | NP_568232.1 | 17 - 18 | 6038 | 67 | |
Set Membership: |
|