Assay Details
| Gene Symbol: AT5G10540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121092.4 | NP_568232.1 | 7 - 8 | 4258 | 97 | |
| GenBank mRNA | AY050978.1 | - | 7 - 8 | 968 | 97 | |
| BT002369.1 | - | 7 - 8 | 951 | 97 | ||
More Information
Set Membership: |
|

AT5G10540
Genomic Map