Assay Details
| Gene Symbol: CM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121125.3 | NP_196648.1 | 2 - 3 | 340 | 148 | |
| GenBank mRNA | AY065238.1 | - | 2 - 3 | 334 | 148 | |
| AY133840.1 | - | 1 - 2 | 236 | 148 | ||
| L47355.1 | - | 2 - 3 | 299 | 148 | ||
More Information
Set Membership: |
|

CM2
Genomic Map