Assay Details
| Gene Symbol: CM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121125.3 | NP_196648.1 | 4 - 5 | 566 | 113 | |
| GenBank mRNA | AY065238.1 | - | 4 - 5 | 560 | 113 | |
| AY133840.1 | - | 3 - 4 | 462 | 113 | ||
| L47355.1 | - | 4 - 5 | 525 | 113 | ||
More Information
Set Membership: |
|

CM2
Genomic Map