Assay Details
| Gene Symbol: CM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121125.3 | NP_196648.1 | 5 - 6 | 662 | 78 | |
| GenBank mRNA | AY065238.1 | - | 5 - 6 | 656 | 78 | |
| AY133840.1 | - | 4 - 5 | 558 | 78 | ||
| L47355.1 | - | 5 - 6 | 621 | 78 | ||
More Information
Set Membership: |
|

CM2
Genomic Map