Assay Details
| Gene Symbol: AT5G10950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121133.4 | NP_196656.1 | 1 - 2 | 274 | 103 | |
| GenBank mRNA | AY063955.1 | - | 1 - 2 | 154 | 103 | |
| AY096725.1 | - | 1 - 2 | 82 | 103 | ||
More Information
Set Membership: |
|

AT5G10950
Genomic Map