Assay Details
| Gene Symbol: AT5G10950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121133.4 | NP_196656.1 | 3 - 4 | 391 | 100 | |
| GenBank mRNA | AY063955.1 | - | 3 - 4 | 271 | 100 | |
| AY096725.1 | - | 3 - 4 | 199 | 100 | ||
More Information
Set Membership: |
|

AT5G10950
Genomic Map