Assay Details
| Gene Symbol: OCP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121165.4 | NP_196688.2 | 2 - 3 | 780 | 69 | |
| GenBank mRNA | BT002017.1 | - | 2 - 3 | 626 | 69 | |
| BT008743.1 | - | 2 - 3 | 619 | 69 | ||
More Information
Set Membership: |
|

OCP3
Genomic Map