Assay Details
| Gene Symbol: CYC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343177.1 | NP_001330176.1 | 8 - 9 | 1070 | 104 | |
| NM_121168.5 | NP_568248.2 | 8 - 9 | 1081 | 104 | ||
| GenBank mRNA | BT026128.1 | - | 7 - 8 | 858 | 104 | |
More Information
Set Membership: |
|

CYC3B
Genomic Map