Assay Details
| Gene Symbol: AT5G11470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343193.1 | NP_001318540.1 | 3 - 4 | 631 | 96 | |
| NM_001343194.1 | NP_001331207.1 | 3 - 4 | 922 | 96 | ||
More Information
Set Membership: |
|

AT5G11470
Genomic Map