Assay Details
| Gene Symbol: MYB3R-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343196.1 | NP_001318541.1 | 3 - 4 | 433 | 121 | |
| NM_001343197.1 | NP_001330337.1 | 3 - 4 | 567 | 121 | ||
| NM_001343198.1 | NP_001330338.1 | 2 - 3 | 234 | 121 | ||
| NM_001343199.1 | NP_001330339.1 | 2 - 3 | 107 | 121 | ||
| NM_121189.4 | NP_568249.1 | 3 - 4 | 494 | 121 | ||
| GenBank mRNA | AF371975.2 | - | 3 - 4 | 434 | 121 | |
| AY519650.1 | - | 2 - 3 | 107 | 121 | ||
More Information
Set Membership: |
|

MYB3R-4
Genomic Map