Assay Details
| Gene Symbol: AT5G11550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121193.3 | NP_196716.1 | 1 | 1199 | 111 | |
| GenBank mRNA | BT002981.1 | - | 1 | 977 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G11550| Gene Symbol: AT5G11550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121193.3 | NP_196716.1 | 1 | 1199 | 111 | |
| GenBank mRNA | BT002981.1 | - | 1 | 977 | 111 | |
Set Membership: |
|