Assay Details
| Gene Symbol: AT5G11900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121228.5 | NP_196751.2 | 2 - 3 | 479 | 87 | |
| GenBank mRNA | AK175446.1 | - | 2 - 3 | 274 | 87 | |
| AK175688.1 | - | 2 - 3 | 413 | 87 | ||
| AK176536.1 | - | 2 - 3 | 313 | 87 | ||
| AK221140.1 | - | 2 - 3 | 384 | 87 | ||
| AY064058.1 | - | 2 - 3 | 384 | 87 | ||
| BT008585.1 | - | 2 - 3 | 244 | 87 | ||
More Information
Set Membership: |
|

AT5G11900
Genomic Map