Assay Details
| Gene Symbol: AT5G11900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121228.5 | NP_196751.2 | 5 - 6 | 685 | 149 | |
| GenBank mRNA | AK175446.1 | - | 5 - 6 | 480 | 149 | |
| AK175688.1 | - | 5 - 6 | 619 | 149 | ||
| AK176536.1 | - | 5 - 6 | 519 | 149 | ||
| AK221140.1 | - | 5 - 6 | 590 | 149 | ||
| AY064058.1 | - | 5 - 6 | 590 | 149 | ||
| BT008585.1 | - | 5 - 6 | 450 | 149 | ||
More Information
Set Membership: |
|

AT5G11900
Genomic Map