Assay Details
| Gene Symbol: PYD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121258.4 | NP_568258.2 | 1 - 2 | 223 | 65 | |
| GenBank mRNA | AF465755.1 | - | 1 - 2 | 84 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PYD2| Gene Symbol: PYD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121258.4 | NP_568258.2 | 1 - 2 | 223 | 65 | |
| GenBank mRNA | AF465755.1 | - | 1 - 2 | 84 | 65 | |
Set Membership: |
|