Assay Details
| Gene Symbol: PYD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121258.4 | NP_568258.2 | 7 - 8 | 1120 | 62 | |
| GenBank mRNA | AF465755.1 | - | 7 - 8 | 981 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PYD2| Gene Symbol: PYD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121258.4 | NP_568258.2 | 7 - 8 | 1120 | 62 | |
| GenBank mRNA | AF465755.1 | - | 7 - 8 | 981 | 62 | |
Set Membership: |
|