Assay Details
| Gene Symbol: PYD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121258.4 | NP_568258.2 | 9 - 10 | 1293 | 75 | |
| GenBank mRNA | AF465755.1 | - | 9 - 10 | 1154 | 75 | |
More Information
Set Membership: |
|

PYD2
Genomic Map