Assay Details
| Gene Symbol: MED19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121261.3 | NP_196784.1 | 4 - 5 | 896 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED19A| Gene Symbol: MED19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121261.3 | NP_196784.1 | 4 - 5 | 896 | 91 | |
Set Membership: |
|