Assay Details
| Gene Symbol: FIS1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121277.4 | NP_568272.1 | 1 - 2 | 294 | 63 | |
| GenBank mRNA | AB195718.1 | - | 1 - 2 | 110 | 63 | |
| AK118241.1 | - | 1 - 2 | 199 | 63 | ||
| BT005496.1 | - | 1 - 2 | 110 | 63 | ||
More Information
Set Membership: |
|

FIS1B
Genomic Map