Assay Details
| Gene Symbol: AFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128015.2 | NP_565572.1 | 1 | 1164 | 138 | |
| GenBank mRNA | AY072009.1 | - | 1 | 1104 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AFR| Gene Symbol: AFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128015.2 | NP_565572.1 | 1 | 1164 | 138 | |
| GenBank mRNA | AY072009.1 | - | 1 | 1104 | 138 | |
Set Membership: |
|