Assay Details
| Gene Symbol: TAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128044.3 | NP_180058.1 | 1 - 2 | 360 | 81 | |
| GenBank mRNA | AK226395.1 | - | 1 - 2 | 360 | 81 | |
| BT002475.1 | - | 1 - 2 | 360 | 81 | ||
| BT006593.1 | - | 1 - 2 | 296 | 81 | ||
More Information
Set Membership: |
|

TAT3
Genomic Map