Assay Details
| Gene Symbol: CLPB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128071.3 | NP_565586.1 | 5 - 6 | 1158 | 70 | |
| GenBank mRNA | AY070722.1 | - | 5 - 6 | 1107 | 70 | |
| BT002223.1 | - | 5 - 6 | 953 | 70 | ||
More Information
Set Membership: |
|

CLPB4
Genomic Map