Assay Details
| Gene Symbol: PKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335976.1 | NP_001324292.1 | 12 - 13 | 1564 | 78 | |
| NM_001335977.1 | NP_001324293.1 | 12 - 13 | 1591 | 78 | ||
| NM_001335978.1 | NP_001324291.1 | 11 - 12 | 1513 | 78 | ||
| NM_128074.4 | NP_565587.1 | 12 - 13 | 1537 | 78 | ||
| GenBank mRNA | AF185577.1 | - | 12 - 13 | 1537 | 78 | |
| AF185578.1 | - | 11 - 12 | 1517 | 78 | ||
More Information
Set Membership: |
|

PKL
Genomic Map