Assay Details
| Gene Symbol: PKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335976.1 | NP_001324292.1 | 21 - 22 | 2699 | 103 | |
| NM_001335977.1 | NP_001324293.1 | 21 - 22 | 2726 | 103 | ||
| NM_001335978.1 | NP_001324291.1 | 20 - 21 | 2648 | 103 | ||
| NM_128074.4 | NP_565587.1 | 21 - 22 | 2672 | 103 | ||
| GenBank mRNA | AF185577.1 | - | 21 - 22 | 2672 | 103 | |
| AF185578.1 | - | 20 - 21 | 2652 | 103 | ||
More Information
Set Membership: |
|

PKL
Genomic Map