Assay Details
| Gene Symbol: PKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335976.1 | NP_001324292.1 | 6 - 7 | 860 | 109 | |
| NM_001335977.1 | NP_001324293.1 | 6 - 7 | 887 | 109 | ||
| NM_001335978.1 | NP_001324291.1 | 5 - 6 | 809 | 109 | ||
| NM_128074.4 | NP_565587.1 | 6 - 7 | 833 | 109 | ||
| GenBank mRNA | AF185577.1 | - | 6 - 7 | 833 | 109 | |
| AF185578.1 | - | 5 - 6 | 813 | 109 | ||
More Information
Set Membership: |
|

PKL
Genomic Map