Assay Details
| Gene Symbol: PKL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335976.1 | NP_001324292.1 | 9 - 10 | 1304 | 132 | |
| NM_001335977.1 | NP_001324293.1 | 9 - 10 | 1331 | 132 | ||
| NM_001335978.1 | NP_001324291.1 | 8 - 9 | 1253 | 132 | ||
| NM_128074.4 | NP_565587.1 | 9 - 10 | 1277 | 132 | ||
| GenBank mRNA | AF185577.1 | - | 9 - 10 | 1277 | 132 | |
| AF185578.1 | - | 8 - 9 | 1257 | 132 | ||
More Information
Set Membership: |
|

PKL
Genomic Map