Assay Details
| Gene Symbol: AT2G25450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128102.6 | NP_180115.1 | 1 - 2 | 592 | 77 | |
| GenBank mRNA | AY050787.1 | - | 1 - 2 | 556 | 77 | |
| AY114055.1 | - | 1 - 2 | 500 | 77 | ||
More Information
Set Membership: |
|

AT2G25450
Genomic Map