Assay Details
| Gene Symbol: emb2410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336007.1 | NP_001324008.1 | 20 - 21 | 6155 | 77 | |
| NM_001336008.1 | NP_001324009.1 | 20 - 21 | 6169 | 77 | ||
| NM_001336009.1 | NP_001324007.1 | 21 - 22 | 5890 | 77 | ||
| NM_128125.3 | NP_180137.3 | 21 - 22 | 5890 | 77 | ||
More Information
Set Membership: |
|

emb2410
Genomic Map