Assay Details
| Gene Symbol: SK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128129.2 | NP_565604.1 | 1 - 2 | 394 | 90 | |
| GenBank mRNA | AY052700.1 | - | 1 - 2 | 394 | 90 | |
| AY139806.1 | - | 1 - 2 | 319 | 90 | ||
More Information
Set Membership: |
|

SK3
Genomic Map