Assay Details
| Gene Symbol: AT2G26230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128180.5 | NP_180191.1 | 5 - 6 | 686 | 76 | |
| GenBank mRNA | AY050318.1 | - | 5 - 6 | 589 | 76 | |
| AY087805.1 | - | 5 - 6 | 639 | 76 | ||
| AY101530.1 | - | 4 - 5 | 489 | 76 | ||
More Information
Set Membership: |
|

AT2G26230
Genomic Map