Assay Details
| Gene Symbol: AT2G26230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128180.5 | NP_180191.1 | 7 - 8 | 953 | 145 | |
| GenBank mRNA | AY050318.1 | - | 7 - 8 | 856 | 145 | |
| AY087805.1 | - | 7 - 8 | 906 | 145 | ||
| AY101530.1 | - | 6 - 7 | 756 | 145 | ||
More Information
Set Membership: |
|

AT2G26230
Genomic Map